missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™

Catalog No. 0666425
Change view
Click to view available options
Conductivity:
10 μ℧
10, 100, 1000, 1413, 10,000, 100,000 μ℧
10,000 μ℧
100 μ℧
100,000 μ℧
1000 μ℧
1413 μ℧
5 μ℧
Quantity:
100 mL
6 x 100 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conductivity Quantity
0666425 5 μ℧ 100 mL
093286 10 μ℧ 100 mL
093287 100 μ℧ 6 x 100 mL
093288 1000 μ℧ 100 mL
0932812 1413 μ℧ 6 x 100 mL
093289 10,000 μ℧ 100 mL
0932810 100,000 μ℧ 100 mL
0932820 10, 100, 1000, 1413, 10,000, 100,000 μ℧ 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 0666425

Ideal for lab or field applications

Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.

Calibration within container — zero contamination

  • Traceable to NIST (National Institute of Standards and Technology)
  • Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
  • 100% compatible with all conductivity meters
  • A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
  • Includes an individual temperature chart and step-by-step calibration instructions
  • Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
  • Single use Eliminates external contamination concerns Each standard contains 100mL

Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12

Order Info

Each bottle contains 100mL

Specifications

Chemical Name or Material Conductivity Standard
Packaging 6/Pk.
Conductivity 5 μ℧
Technique Calibration of Conductivity Instrument
Color Clear
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 100 mL