missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Datalogging Thermometer
Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.
Description
Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.
- Data-logging thermometer with fixed one-minute timing intervals
- Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
- Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
- Stores up to ten unique alarm events
- Capable of storing over 1 million temperature observations
- Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
- Status indicators: Low battery, low memory, USB data transfer and active alarm state
- Traceable to NIST for accuracy
- High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
- Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
- Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
- Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
- Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
- All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
- No software is needed for computer interface
- Each thermometer has a unique ID allowing multiple units to be used in the same location
- Raw data on unit cannot be overwritten or modified
- Indicator light will appear when 95% of memory is full
Data-Logging Features
- Can be used in environments requiring 21 CFR 11 compliance
- Thermometer logs two years of observations with a one-minute logging interval
- Device will stop recording when storage is full and will not overwrite recorded data
- Data is unable to be deleted from memory once stored
Bottle Probe Features
- Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
- Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
- Eliminates concerns about incidental contact with food or drinking water
- Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
- Ideal for use in monitoring storage conditions for vaccines or other important samples
- Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program
Order Info
Replacement battery: Cat. No. 1111
Chemical Identifiers
| Datalogger | |
| NIST, ISO/IEC 17025, A2LA, CDC | |
| 0.01° | |
| -58°F to +158°F | |
| 5 oz. |
| TraceableLIVE | |
| ±0.25°C | |
| -50°C to +70°C | |
| -72.4°F to +316.4°F | |
| 141.7 g |
Specifications
| Datalogger | |
| Not Calibrated | |
| TraceableLIVE | |
| 2 Probes, 5/16 in. dia. x 3/4 in. long | |
| USB | |
| ±0.25°C | |
| -50°C to +70°C | |
| -72.4°F to +316.4°F | |
| 141.7 g |
| LCD | |
| LED Indicator, Audible, Flashing LCD Segment | |
| 2 Probes, 5/16 in. dia. x 3/4 in. long | |
| NIST, ISO/IEC 17025, A2LA, CDC | |
| Temperature-Buffered Bottle Sensor | |
| 0.01° | |
| -58°F to +158°F | |
| 5 oz. |