missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals - Loamy Sand 4, MilliporeSigma™ Supelco™

Catalog No. 111016957
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016957 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016957 Supplier Merck Emd Millipore Supplier No. CRM03650G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 1310-58-3
pH 4.34
Type Certified Reference Material
Quantity 50 g
Grade Certified Reference Material