missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals 3 For Environmental Analysis, SPEX CertiPrep™

Catalog No. MN6
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MN6 125 mL
MN6500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MN6 Supplier SPEX CertiPrep Group LLC Supplier No. MN6

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba500μg/mL
Ca500μg/mL
Mo500μg/mL
Na500μg/mL
K100μg/mL
Mg100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Trace Metals Standard 3
Packaging Triple-leached LDPE bottle
Name Note Groundwater and Wastewater Pollution Control Check Standard
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Industry Type Wastewater Testing
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Ba    •500μg/mL
Ca    •500μg/mL
Mg    •100μg/mL
Mo    •500μg/mL
K    •100μg/mL
Na    •500μg/mL
Show More Show Less

Product intended for laboratory use only