missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Metals 1 without Mercury For Environmental Analysis, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Al | 500μg/mL |
| V | 250μg/mL |
| As | 100μg/mL |
| Be | 100μg/mL |
| Co | 100μg/mL |
| Cr | 100μg/mL |
| Cu | 100μg/mL |
| Fe | 100μg/mL |
| Mn | 100μg/mL |
| Ni | 100μg/mL |
| Pb | 100μg/mL |
| Zn | 100μg/mL |
| Cd | 25μg/mL |
| Se | 25μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Trace Metals I without Mercury |
| Packaging | Triple-leached LDPE bottle |
| Name Note | Groundwater and Wastewater Pollution Control Check Standard |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | 5% HNO3 |
| Industry Type | Wastewater Testing |
| Quantity | 125 mL |
| Grade | Environmental |
| Show More |
Product intended for laboratory use only