missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Metals 2 - WP, Certified Refernce Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 7697-37-2 |
| Type | Certified Reference Material |
| Quantity | 20 mL |
| Grade | Certified Reference Material |