missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60)

Catalog No. PIMA1023X
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1023X 20 μL
PIMA1023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA1023X Supplier Invitrogen™ Supplier No. MA1023X
Only null left

Mouse Monoclonal Antibody

TRA-1-60 Monoclonal Antibody for Flow, ICC/IF, Western Blot

TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.

Specifications

Antigen TRA-1-60
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TRA-1-60
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Alias TRA160; tra60
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep.
Purification Method Affinity chromatography - MBP
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less