missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging
Description
The Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits are the only kits offering superior imaging for live human PSCs in one box. This TRA-1-60 Alexa Fluor™ 555 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.
With the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging, you can:
Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM imaging medium
Stain live cell samples with confidence using reagents that are tested to be free of common contaminants
Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM imaging medium.
Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM imaging medium maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.
Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2μm sterile-filtered and tested to be free of common contaminants.
What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 uL staining volume and includes:
200μL of Alexa Fluor™ 555 Mouse Anti-Human TRA-1-60 (50X concentration)
500 mL FluoroBrite™ DMEM
With the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging, you can:
Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM imaging medium.
Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM imaging medium maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.
Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2μm sterile-filtered and tested to be free of common contaminants.
What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 uL staining volume and includes:
Specifications
Specifications
| Conjugate | Alexa Fluor 555 |
| Quantity | 50 tests |
| Content And Storage |
|
| Product Type | Conjugate Kit |
For Research Use Only. Not for use in diagnostic procedures.