missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRA-1-60 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging

Catalog No. A25618
Encompass
Change view
Click to view available options
Quantity:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25618 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25618 Supplier Invitrogen™ Supplier No. A25618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits provide reagents for superior imaging of live human PSCs.

This TRA-1-60 Alexa Fluor™ 488 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers and germ layer markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.

With the TRA-1-60 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging, you can:
• Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
• Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM
• Stain live cell samples with confidence using reagents that are tested to be free of common contaminants

Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM.

Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.

Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2 μm sterile-filtered and tested to be free of common contaminants.

What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 488 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 μL staining volume and includes:
• 200 μL of Alexa Fluor™ 488 Mouse Anti-Human TRA-1-60 (50X concentration)
• 500 mL FluoroBrite™ DMEM

Specifications

Conjugate Alexa Fluor 488
Quantity 50 tests
Content And Storage
  • 200 μL of Alexa Fluor™ 488 Mouse Anti-Human TRA-1-60 (50 x concentration) (store at 2°C to 8°C)
  • 500 mL FluoroBrite™ DMEM
  • Store at 2°C to 8°C, protect from light
)
Product Line Alexa Fluor
Product Type Conjugate Kit

For Research Use Only. Not for use in diagnostic procedures.