missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPRG1 Polyclonal Antibody

Description
Immunogen sequence: TFTEHPMKYT SEKFLEICKL SGFMSKLVPA IQNAHKNSTG SGRGKKLMVL TEPILIETYT G Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
Specifications
Specifications
| Antigen | TPRG1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TPRG1 |
| Gene Accession No. | Q6ZUI0 |
| Gene Alias | FAM79B; family with sequence similarity 79, member B; Protein FAM79B; TPRG1; tumor protein p63 regulated 1; tumor protein p63-regulated gene 1 protein |
| Gene Symbols | TPRG1 |
| Show More |