missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
This gene encodes a large coiled-coil protein that forms intranuclear filaments attached to the inner surface of nuclear pore complexes (NPCs). The protein directly interacts with several components of the NPC. It is required for the nuclear export of mRNAs and some proteins. Oncogenic fusions of the 5′ end of this gene with several different kinase genes occur in some neoplasias.
Specifications
Specifications
| Antigen | TPR |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | TPR |
| Gene Accession No. | P12270 |
| Gene Alias | translocated promoter region, nuclear basket protein |
| Gene Symbols | TPR |
| Show More |