Learn More
TPH in Water (Low Level), Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017302
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
111017302 2 mL
1 options
Catalog No. 111017302 Supplier Merck Emd Millipore Supplier No. PE17992ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This vial contains a concentrate of a blended gasoline from three different sources and diesel fuel oil No. 2 in pentane for analysis by TNRCC Method 1005. When prepared according to instructions, the concentration of each analyte will be less than 10 mg/L. Sample preparationTo prepare solution, fill a 40-mL VOA vial to volume with carbon-filtered reagent-grade water at room temperature. Remove enough water to allow for the addition of extraction solvent. Open vial PE1799-2ML and immediately withdraw 40 muL with a gas tight syringe. Quickly inject the 40 muL aliquot below the surface of the reagent water in the voa vial. Add extraction solvent. Mix the sample by inverting the flask three times only (excessive shaking results in loss of volatiles) and analyze immediately. Assume a 40 mL sample for calculation purposes. Note this is a 1:1000 dilution and other dilutions are acceptable.
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) Residual Range Organics (RRO),>C28-C35 0.000 - 10.0 mg/L; Total Petroleum Hydrocarbons (TPH), (C6-C35 5-50 mg/L; Diesel-range total petroleum hydrocarbons, >C12-C28 0.000 - 10.0 mg/L; Gasoline Range Organics, C6-C12 0.000 - 10.0 mg/L)