missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPH - Sandy Loam 1, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Soil |
| CAS | 64742-65-0 |
| pH | 7.51 |
| Type | Certified Reference Material |
| Quantity | 100 g |
| Grade | Certified Reference Material |