missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPH Mix Certified Reference Material, 2,000 μg/mL in Methylene Chloride, MilliporeSigma™ Supelco™
Specifications
Specifications
| Concentration | 2,000 μg/mL in dichloromethane: carbon disulfide (50:50) |
| Quantity | 1.5 mL |
| Grade | Certified Reference Material |
| Concentration or Composition (by Analyte or Components) | Decane; Docosane; Dodecane; Dotriacontane; Eicosane; Hexacosane; Hexadecane; Hexatriacontane; Octacosane; Octadecane; Octane; Octatriacontane; Tetracontane; Tetracosane; Tetradecane; Tetratriacontane; Triacontane |