missing translation for 'onlineSavingsMsg'
Learn More

TPH -Loamy Sand 1, MilliporeSigma™ Supelco™

Catalog No. 111001708
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001708 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001708 Supplier Merck Emd Millipore Supplier No. CRM352100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TPH Source - Light Refinery Oil

Specifications

CAS 64742-65-0
pH 7.50
Synonym Total Petroleum Hydrocarbon - Loamy Sand 1
Type Certified Reference Material
Quantity 100 g
Grade Certified Reference Material