missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53I11 Polyclonal Antibody
TP53I11 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA563940
Description
TP53I11 Polyclonal Antibody for ICC/IF, IHC
Specifications
| TP53I11 | |
| Polyclonal | |
| Unconjugated | |
| TP53I11 | |
| p53-induced gene 11 protein; Pig11; TP53I11; transformation related protein 53 inducible protein 11; Trp53i11; tumor protein p53 inducible protein 11; tumor protein p53-inducible protein 11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9537 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14683 | |
| TP53I11 | |
| Recombinant protein corresponding to Human TP53I11. Recombinant protein control fragment (Product #RP-101958). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |