missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TP53I11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116061
Description
Antibody detects endogenous levels of total TP53I11.
TP53I11 is a 177 amino acid tumor suppressor belonging to the p53-induced protein gene (PIG) family. The PIG gene family encodes redox-controlling proteins that are involved in p53 tumor suppressor activity. It is suggested that PIG11 is involved in arsenic trioxide As(2)O(3)-induced apoptosis in certain cell lines and may play a significant role in tumor suppression through promotion of cell apoptosis. The gene encoding PIG11 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome.Spécifications
| TP53I11 | |
| Polyclonal | |
| Unconjugated | |
| TP53I11 | |
| p53-induced gene 11 protein; Pig11; TP53I11; transformation related protein 53 inducible protein 11; Trp53i11; tumor protein p53 inducible protein 11; tumor protein p53-inducible protein 11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9537 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O14683 | |
| TP53I11 | |
| A synthesized peptide derived from human TP53I11(Accession O14683), corresponding to amino acid residues L9-P59. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |