missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ToxBLAzer™ DualScreen Kit

Catalog No. p-7076303
Encompass
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1138 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1138 Supplier Invitrogen™ Supplier No. K1138
Only null left

ToxBLAzer™ DualScreen Kit

ToxBLAzer™ DualScreen Kit provide the key advantage of combining the ratiometric reporter gene readout of beta-lactamase with a cytotoxicity readout in the same assay. Thus, ToxBLAzer™ DualScreen yields reporter readouts that normalize cytotoxic cellular responses, which is a necessity in screening applications which down regulate transcription. The ToxBLAzer™ substrate contains both the LiveBLAzer™-FRET B/G substrate and a proprietary cytotoxicity indicator. Both compounds readily enter the cell, where endogenous esterases rapidly convert them into fluorescent compounds Esterase activity and an intact cell membrane are required for detection of the indicator. The cytotoxicity indicator's fluorescence is red-shifted to ensure compatibility with the blue and green emissions of the LiveBLAzer™-FRET B/G substrate, allowing detection of both reporter gene activity and cytotoxicity in the same assay sample.

Advantages of ToxBLAzer™ DualScreen Kits:

  • Multiplexed readout results in rapid identification of false positives due to toxicity or lack of viable cells, thereby improving hit-picking efficiency
  • Ratiometric blue:green reporter readout reduces false hits by minimizing experimental noise
  • Single reagent addition eliminates need for follow up cytotoxicity assays, thereby increasing throughput while reducing sample and test compound consumption
  • Specifications

    Product Line ToxBLAzer
    For Use With (Equipment) Microplate Reader, Fluorometer, Flow Cytometer
    Content And Storage ToxBLAzer™ DualScreen Loading Kits include ToxBLAzer™ substrate, DMSO (for Solution A), Solution B, and Solution C. ToxBLAzer™ substrate is provided lyophilized and should be stored at 20°C, dessicated and protected from light. DMSO (for Solution A), Solution B, and Solution C should be stored at 22-25°C, protected from direct light.

    The 200 μg kit is sufficient for 3,750 x 48 μl standard protocol well assays; the 5 mg kit is sufficient for 93,750 x 48 μl standard protocol well assays. Bulk quantities of loading kits are available upon request.
    Detection Method Fluorescence
    Product Type Dual Screen Kit
    Quantity 200 μg
    Format Kit
    Cell Permeability Cell-Permeant
    Substrate Beta-Lactamase Substrate

    For Research Use Only. Not for use in diagnostic procedures.