missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughWash Washdown Resistant Labels for BJet J4000 Printer

Catalog No. 17378732
Change view
Click to view available options
Width (English):
6 in.
7 in.
8 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
17378732 7 in.
17378731 6 in.
17378727 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17378732 Supplier Brady™ Supplier No. J40C70002855

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sets the standard for durability and vibrant full-color identification in harsh washdown environments.

  • Labels and ink withstand high-pressure, high-temperature water as well as chemical exposure and scrubbing
  • Create custom ANSI and OSHA header signage in any color and multiple languages
  • Print-on-demand capabilities eliminate the need to order and stock pre-printed labels
  • Enhanced label topcoat works with durable J4000 inks to protect printed text and graphics from smudging
  • Service temperature range of -40 to 140°F
  • An ink cartridge is required to print this material use part number J40-CMY

Specifications

Adhesive Type Permanent Acrylic
Color White
Finish Matte
Format Roll
Material Inkjet-receptive Polyester
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
Length (English) 100 ft.
Width (English) 7 in.