Learn More
Ansell TouchNTuff™ 83-500 Sterile Polyisoprene Gloves
Sterile polyisoprene gloves are ergonomically-designed to enhance fit and comfort, and reduce hand fatigue and stress.
Supplier:  Ansell 83500070
 View more versions of this product
Catalog No. 17000837
Only null left
Explore available promotions
Description
Description
  • Powder-free, chlorinated glove interior
  • Rolled bead cuff style
  • Hand specific
  • Smooth finish
  • Polyisoprene
  • White
Specifications
Specifications
Class 100 (ISO 5)
0.65
Single-use
Specific
Polyisoprene
Cleanroom Gloves
Sterile
Beaded/Rolled
200/Pk.
83-500
300
No
White
No Texture
Yes
Powder Free
7
0.21 mm
83-500_7
Splash resistance
12
Product Suggestions
Product Suggestions