missing translation for 'onlineSavingsMsg'
Learn More

Total Residual Chlorine (Low Level) - WP, MilliporeSigma™ Supelco™

Catalog No. 111009344
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111009344 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009344 Supplier Merck Emd Millipore Supplier No. 568573U

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Specifications

Packaging Ampule
Type Proficiency Testing Material
Quantity 2 mL