missing translation for 'onlineSavingsMsg'
Learn More

Total Ionic Strength Adjustment Buffer (TISAB IV), with Tartrate, Ricca Chemical

Catalog No. 867316
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
867316 500 mL
867332 1 L
86731 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 867316 Supplier Ricca Chemical Company Supplier No. 867316
Only null left

For Analysis of Fluoride Analysis Using Ion Selective Electrodes

Specifications

Chemical Name or Material Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB IV)
Name Note With Tartrate
CAS 7732-18-5, 77-86-1, 6106-24-7, 7647-01-0
CAS Min % 62.0, 15.1, 14.35, 6.18
Color Yellow to Green
CAS Max % 65.0, 15.72, 14.94, 6.43
Concentration 1X
For Use With (Application) Fluoride Analysis
For Use With (Equipment) pH Meter and Electrode
pH 9
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less