missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II) |
| Name Note | With CDTA |
| CAS | 7732-18-5, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7 |
| CAS Min % | 82.0, 5.87, 4.91, 4.77, 0.33 |
| CAS Max % | 85.0, 6.11, 5.12, 4.96, 0.34 |
| Concentration | 1X |
| For Use With (Application) | Fluoride Analysis |
| For Use With (Equipment) | Ion Selective Electrode |
| pH | 5.4 |
| Physical Form | Liquid |
| Show More |