missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toremifene Citrate 98.0+%, TCI America™
$288.94 - $866.79
Chemical Identifiers
| CAS | 89778-27-8 |
|---|---|
| Molecular Formula | C32H36ClNO8 |
| Molecular Weight (g/mol) | 598.089 |
| MDL Number | MFCD01729016 |
| InChI Key | IWEQQRMGNVVKQW-OQKDUQJOSA-N |
| Synonym | (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate |
| PubChem CID | 3005572 |
| IUPAC Name | 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
| SMILES | CN(C)CCOC1=CC=C(C=C1)C(=C(CCCl)C2=CC=CC=C2)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
Chemical Identifiers
| 89778-27-8 | |
| 598.089 | |
| IWEQQRMGNVVKQW-OQKDUQJOSA-N | |
| 3005572 | |
| CN(C)CCOC1=CC=C(C=C1)C(=C(CCCl)C2=CC=CC=C2)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
| C32H36ClNO8 | |
| MFCD01729016 | |
| (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate | |
| 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
Specifications
| 89778-27-8 | |
| White-Yellow | |
| MFCD01729016 | |
| (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate | |
| CN(C)CCOC1=CC=C(C=C1)C(=C(CCCl)C2=CC=CC=C2)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O | |
| 598.089 | |
| 598.09 | |
| Crystalline Powder |
| 162°C | |
| C32H36ClNO8 | |
| 1 g | |
| IWEQQRMGNVVKQW-OQKDUQJOSA-N | |
| 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 3005572 | |
| ≥98.0% (HPLC,T) | |
| Toremifene Citrate |
Safety and Handling
RTECSNumber : KH2156700
TSCA : No
Recommended Storage : Refrigerator