CAS RN 89778-27-8
CAS RN 89778-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Toremifene Citrate 98.0+%, TCI America™
CAS: 89778-27-8 Molecular Formula: C32H36ClNO8 Molecular Weight (g/mol): 598.089 MDL Number: MFCD01729016 InChI Key: IWEQQRMGNVVKQW-OQKDUQJOSA-N Synonym: (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate PubChem CID: 3005572 IUPAC Name: 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: CN(C)CCOC1=CC=C(C=C1)C(=C(CCCl)C2=CC=CC=C2)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O