missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Toremifene, 98+%
A chlorinated tamoxifen analogue that induces apoptosis in some cells | CAS: 89778-26-7 | C26H28ClNO, C26H28ClNO | 405.97 g/mol
$221.10 - $386.49
Chemical Identifiers
| CAS | 89778-26-7 |
|---|---|
| Molecular Formula | C26H28ClNO, C26H28ClNO |
| Synonym | Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ63803ME
|
Thermo Scientific Chemicals
J63803ME |
500 mg |
Each for $221.10
|
|
|||||
|
AAJ6380303
|
Thermo Scientific Chemicals
J6380303 |
1 g |
Each for $386.49
|
|
|||||
Description
A chlorinated tamoxifen analogue, that induces apoptosis in some cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89778-26-7 | |
| C26H28ClNO, C26H28ClNO | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine |
Specifications
| White to Off-white | |
| C26H28ClNO, C26H28ClNO | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine | |
| Apoptosis Reagent | |
| A chlorinated tamoxifen analogue that induces apoptosis in some cells | |
| CN(C)CCOC1=CC=C(C=C1)C(=C(\CCCl)C1=CC=CC=C1)\C1=CC=CC=C1 | |
| 405.97 | |
| ≥98% |
| 89778-26-7 | |
| 405.96g/mol | |
| 108°C to 110°C | |
| MFCD00801070 | |
| XFCLJVABOIYOMF-QPLCGJKRSA-N | |
| (2-{4-[(1Z)-4-chloro-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine | |
| Powder |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only