missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toremifene, 98+%

Catalog No. AAJ6380303
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6380303 1 g
AAJ63803ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6380303 Supplier Thermo Scientific Chemicals Supplier No. J6380303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A chlorinated tamoxifen analogue that induces apoptosis in some cells | CAS: 89778-26-7 | C26H28ClNO, C26H28ClNO | 405.97 g/mol

A chlorinated tamoxifen analogue, that induces apoptosis in some cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89778-26-7
Molecular Formula C26H28ClNO
Synonym Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine

Specifications

Description White to Off-white
Quantity 1 g
Formula Weight 405.96g/mol
Melting Point 108°C to 110°C
Product Type Apoptosis Reagent
For Use With (Application) A chlorinated tamoxifen analogue that induces apoptosis in some cells
Form Powder
Assay ≥98%

RUO – Research Use Only