missing translation for 'onlineSavingsMsg'
Learn More

Topiramate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012250
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012250 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012250 Supplier Merck Emd Millipore Supplier No. PHR16001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Topiramate, a sulfamate-substituted derivative of the monosaccharide D-fructose , is a medication, which is widely used for the treatment of epilepsy.

Specifications

CAS 97240-79-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H21 NO8 S
MDL Number MFCD00865320
Synonym Topiramate; 2,3:4,5-Bis-O-(1-methylethylidene)-36-D-fructo-pyranose sulfamate; McN 4853; RWJ 17021; Topamax
Type Certified Reference Material
Quantity 1 g
Formula Weight 339.36
Grade Certified Reference Material