Learn More
MSA™ Topgard™ Protective Caps
Designed for use by utilities and for the environments where elevated temperature hazards are present
$48.63 - $60.69
| Catalog Number | Mfr. No. | Color | Suspension Type | Head Style | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Suspension Type | Head Style | Price | Quantity | |||
|
19084104
|
MSA™
454728 |
White | 1-Touch | Slotted |
Each for $48.63
|
|
|||
|
18-999-3796
|
MSA™
475380 |
Blue | Fas-Trac Ratchet | Slotted |
Each for $60.69
|
|
|||
Description
- Lightweight, durable, slotted cap
- Shell constructed of impact-resistant, injection-molded polycarbonate
- Withstands radiant heat loads up to, but not exceeding, 275°F
- 1-Touch™ suspension with one-handed slide adjustment and an easy squeeze opening
- Four-point webbed Fas-Trac™ ratchet suspension with vinyl sweatband and large easy-adjustment knob
Compliance
Third-party ceritfied by SEI to meet Type I, Class E requirements for ANSI/ISEA Z89.1 and CSA Z94.1.
Specifications
| Polycarbonate |