Learn More
MSA™ Topgard™ Protective Caps
Designed for use by utilities and for the environments where elevated temperature hazards are present
Supplier: MSA™ 475380
Description
- Lightweight, durable, slotted cap
- Shell constructed of impact-resistant, injection-molded polycarbonate
- Withstands radiant heat loads up to, but not exceeding, 275°F
- 1-Touch™ suspension with one-handed slide adjustment and an easy squeeze opening
- Four-point webbed Fas-Trac™ ratchet suspension with vinyl sweatband and large easy-adjustment knob
Compliance
Third-party ceritfied by SEI to meet Type I, Class E requirements for ANSI/ISEA Z89.1 and CSA Z94.1.
Specifications
| Meets ANSI Z89.1-2003; Type I; Class E and CSA Z94.1-2005. | |
| Polycarbonate | |
| Slotted |
| Blue | |
| Fas-Trac Ratchet |