missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1 Recombinant Human Monoclonal Antibody (K94015_6C5), Invitrogen™
Human Recombinant Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | TOP1 |
|---|---|
| Clone | K94015_6C5 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
Description
Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI
TOP1 Recombinant Monoclonal antibody specifically detects TOP1 in Human samples. It is validated for ELISA, Western BlotSpecifications
| TOP1 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase | |
| Recombinant human TOP1 (immunogen range is 221-420). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Top1 |
| K94015_6C5 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Human | |
| Human | |
| P11387 | |
| 7150 | |
| IgG1 | |
| Protein A | |
| Antibody |