missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Top Dog 8 mil Die Cut 50 in. Apron

Top Dog 8 mil Die Cut 50 in. Apron is reversable and designed to fit comfortably over clothes.

Supplier:  Vikan™ 81025

 View more versions of this product

Catalog No. 17-227-955


Only null left
Explore available promotions

Description

Description

  • Reusable
  • Color-coded
  • FDA-compliant garments
  • Designed to support zoned color-coding programs
  • Thicker 8 mil material significantly increases resistance to punctures and tears
  • Ideal for workstations with sharp edges or jobs that require lifting, carrying, or the use of knives or scissors
  • One-piece construction limits the potential for foreign body contamination and reduces harborage points for pathogens
Specifications

Specifications

Apron
Polyurethane
Reusable
Non-sterile
8 mil
50 x 50 in.
0.6 lb.
Universal
White
Waist Ties
Ether-Based Polyurethane
FDA Compliant
50 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions