missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOMM20 Monoclonal Antibody (K16238_5F9), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555630

 View more versions of this product

Catalog No. MA555630


Only null left
Explore available promotions

Description

Description

The mitochondrial preprotein translocases of the outer membrane (Tom) is a multisubunit protein complex that facilitates the import of nucleus-encoded precursor proteins across the mitochondrial outer membrane. The Tom machinery consists of import receptors for the initial binding of cytosolically synthesized preproteins and a general import pore (GIP) for the membrane translocation of various preproteins into the mitochondria. The import receptors include Tom20 and Tom22, which form a heteromeric receptor complex that initiates the insertion of newly synthesized proteins into the outer membrane and then directs the precursor protein into the GIP. In yeast, Tom22 is the essential component of the import receptor complex as it functions as both a receptor for the preproteins and serves as a docking point for both Tom20 and the GIP.
Specifications

Specifications

TOMM20
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q15388, Q62760, Q9DCC8
TOMM20
Recombinant human TOMM20 (immunogen range is 31-140).
50 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
K16238_5F9
Unconjugated
TOMM20
1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; mKIAA0016; MOM19; outer mitochondrial membrane receptor rTOM20; outer mitochondrial membrane receptor Tom20; TOM20; TOMM20; translocase of outer mitochondrial membrane 20; translocase of outer mitochondrial membrane 20 homolog (yeast); translocase of outer mitochondrial membrane 20 homolog type II
Mouse
Protein A
RUO
266601, 67952, 9804
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions