missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolylene-2,6-diisocyanate 98.0+%, TCI America™
Supplier: TCI America T11535G
Specifications
| Tolylene-2,6-diisocyanate | |
| Yellow | |
| C9H6N2O2 | |
| 5 g | |
| 2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene | |
| CC1=C(C=CC=C1N=C=O)N=C=O | |
| 174.159 | |
| CHEBI:53557 | |
| ≥98.0% (GC) |
| 91-08-7 | |
| 103°C | |
| MFCD00002010 | |
| 2078 | |
| RUELTTOHQODFPA-UHFFFAOYSA-N | |
| 1,3-diisocyanato-2-methylbenzene | |
| 7040 | |
| 174.16 | |
| Liquid |
Chemical Identifiers
| 91-08-7 | |
| 174.159 | |
| RUELTTOHQODFPA-UHFFFAOYSA-N | |
| 7040 | |
| 1,3-diisocyanato-2-methylbenzene |
| C9H6N2O2 | |
| MFCD00002010 | |
| 2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene | |
| CHEBI:53557 | |
| CC1=C(C=CC=C1N=C=O)N=C=O |
Safety and Handling
EINECSNumber : (3)-2214
RTECSNumber : CZ6310000
TSCA : Yes
Recommended Storage : Refrigerator