missing translation for 'onlineSavingsMsg'
Learn More

Tolylene-2,6-diisocyanate 98.0+%, TCI America™

Catalog No. T11531G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11531G 1 g
T11535G 5 g
T115325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T11531G Supplier TCI America Supplier No. T11531G

Chemical Identifiers

CAS 91-08-7
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00002010
InChI Key RUELTTOHQODFPA-UHFFFAOYSA-N
Synonym 2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
PubChem CID 7040
ChEBI CHEBI:53557
IUPAC Name 1,3-diisocyanato-2-methylbenzene
SMILES CC1=C(C=CC=C1N=C=O)N=C=O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 1 g
UN Number 2078
Formula Weight 174.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tolylene-2,6-diisocyanate
EINECSNumber (3)-2214
RTECSNumber CZ6310000
TSCA Yes
Recommended Storage Refrigerator