missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluidine Blue Stain, 1% (w/v), Ricca Chemical
Specifications
Specifications
| CAS Min % | 97.0, 0.97 |
| CAS Max % | 100.0, 1.01 |
| Concentration | 1% (w/v) |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Blue/Purple |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Percent Purity | 1% W/V |
| Grade | Laboratory |
| Show More |