missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluidine Blue, 1% Aqueous, Certified, LabChem™
Specifications
Specifications
| Absorbance | Passes Test |
| CAS Max % | 99, 1 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Linear Formula | C15H16ClN3S |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds |
| Color | Purple |
| Solubility Information | Soluble in water |
| Quantity | 4 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |