missing translation for 'onlineSavingsMsg'
Learn More

Toluidine Blue, 1% Aqueous, Certified, LabChem™

Catalog No. LC261651
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC261651 500 mL Poly Bottle
LC261652 1 L Poly Bottle
LC261654 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC261651 Supplier LabChem Supplier No. LC261651
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-31-9 , 7732-18-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
PubChem CID 7083
IUPAC Name 3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Absorbance Passes Test
CAS Max % 99, 1
Identification Passes Test
Packaging Poly Bottle
Linear Formula C15H16ClN3S
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Purple
Solubility Information Soluble in water
Quantity 500 mL
Physical Form Liquid
Formula Weight 305.82
Grade Certified
Chemical Name or Material Toluidine Blue, 1% Aqueous
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature