missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.7% (GC)
Supplier: Honeywell Inc 322494X25L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Toluene | |
| Reag. Ph. Eur. reag. ISO | |
| Colorless liquid | |
| 110°C to 111°C | |
| 0.07 | |
| 997°F | |
| n20/D 1.496 to 1.498 | |
| Free acid: ≤0.001% (as HCl) | |
| Free alkali: ≤0.001% (as NAOH) | |
| MFCD00008512 | |
| UN1294 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CC1=CC=CC=C1 | |
| 92.14 | |
| CHEBI:17578 | |
| ≥99.7% (GC) |
| -93°C | |
| 108-88-3 | |
| 0.865g/mL at 25°C | |
| 4 x 2.5 L | |
| 4°C (39.2°F) | |
| Glass Bottle | |
| 0 to 0.001% | |
| C7H8 | |
| C6H5CH3 | |
| 22mmHg (20°C), 26mmHg (25°C) | |
| 635760 | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| toluene | |
| 1140 | |
| 92.14g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 108-88-3 | |
| 92.14 | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| 1140 | |
| toluene |
| C7H8 | |
| MFCD00008512 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CHEBI:17578 | |
| CC1=CC=CC=C1 |
Safety and Handling
P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
H225-H304-H315-H336-H361d-H373
Danger
DOTInformation : Class 3 - PG 2 - Toluene
EINECSNumber : 203-625-9
RTECSNumber : XS5250000