missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Solstice

Catalog No. 60046101
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046101 2.5 L Glass Bottle
60046105 6 x 1 L Glass Bottle
60046103 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046101 Supplier Honeywell Inc Supplier No. 322492.5L

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -93°C
Appearance Colorless liquid
Density 0.865g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Packaging Glass Bottle
Refractive Index n20/D 1.496 to 1.498
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid: ≤0.001% (as HCl)
Titratable Base Free alkali: ≤0.001% (as NAOH)
Vapor Pressure 22mmHg (20°C), 26mmHg (25°C)
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110°C to 111°C
Formula Weight 92.14g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Toluene, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361d-H373
DOTInformation Class 3 - PG 2 - Toluene
EINECSNumber 203-625-9
RTECSNumber XS5250000