missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Purified, Reagents

Catalog No. 01336927
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336927 200 L
01336926 20 L
01336925 4 L
01336924 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336927 Supplier Labchem Inc Supplier No. C2341000200E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Toluene, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym Methylbenzene, Phenylmethane, Toluol
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.866
Boiling Point 111°C
Grade Pure
Chemical Name or Material Toluene, Purified