missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.8%, Solstice

Catalog No. 6001726
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001726 204 L Metal Drum
6001725 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001726 Supplier Solstice Supplier No. PP347204

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

CAS Max % 100.0000%
Density 0.867 g/cm3
Quantity 204 L
Flash Point 4°C
Packaging Metal Drum
Water 0.03% max.
UN Number UN1294
Beilstein 635760
Boiling Point 110.6°C
Solubility Information 5.2% in Water at 20°C
Formula Weight 92.14
Percent Purity >99.8%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Toluene, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene