missing translation for 'onlineSavingsMsg'
Learn More

Toluene For spectroscopy Uvasol™, MilliporeSigma™

Catalog No. M1083311000
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1083311000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1083311000 Supplier Technidata Supplier No. 1.08331.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For spectroscopy Uvasol™

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87g/cm3 at 20°C
Quantity 1 L
Flash Point 4.4°C (39.9°F), Closed cup
Ignition Point 535°C (995°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.49
Vapor Pressure 29 hPa (20°C)
UN Number 1294
Boiling Point 110.6°C
Solubility Information Soluble in water: 0.52g/L at 16°C
Formula Weight 92.14
Chemical Name or Material Toluene
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.