missing translation for 'onlineSavingsMsg'
Learn More

Toluene [for Spectrophotometry], TCI America™

Catalog No. T0260100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0260100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0260100ML Supplier TCI America Supplier No. T0260100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Quantity 100 mL
Physical Form Clear Liquid at 20°C
UN Number 1294
Boiling Point 110°C
Percent Purity ≥99.5% (GC)
Chemical Name or Material Toluene [for Spectrophotometry]
EINECSNumber (3)-0002&(3)-0060
RTECSNumber XS5250000
TSCA Yes