missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene For gas chromatography MS SupraSolv™, MilliporeSigma™
For gas chromatography MS SupraSolv™
$239.61 - $500.17
Chemical Identifiers
| CAS | 108-88-3 |
|---|---|
| Molecular Formula | C7H8 |
| Molecular Weight (g/mol) | 92.14 |
| MDL Number | MFCD00008512 |
| InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
| Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
| PubChem CID | 1140 |
| ChEBI | CHEBI:17578 |
| IUPAC Name | toluene |
| SMILES | CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1008491000
|
Technidata
1008491000 |
1 L | Glass Bottle |
Each for $239.61
|
|
||||
|
M1008492500
|
Technidata
1008492500 |
2.5 L | Glass Bottle |
Each for $500.17
|
|
||||
Chemical Identifiers
| 108-88-3 | |
| 92.14 | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| 1140 | |
| toluene |
| C7H8 | |
| MFCD00008512 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CHEBI:17578 | |
| CC1=CC=CC=C1 |
Specifications
| -95°C | |
| Colorless | |
| 110.6°C | |
| 4.4°C (39.9°F), Closed cup | |
| Charactristic | |
| Liquid | |
| C7H8 | |
| 29 hPa (20°C) | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| toluene | |
| 1140 | |
| 92.14 |
| 108-88-3 | |
| 0.87g/cm3 at 20°C | |
| 1 L | |
| 535°C (995°F) | |
| Glass Bottle | |
| 1.49 | |
| MFCD00008512 | |
| 1294 | |
| Soluble in water: 0.52g/L at 16°C | |
| CC1=CC=CC=C1 | |
| 92.14 | |
| CHEBI:17578 | |
| Toluene |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H315: Causes skin irritation.
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.