missing translation for 'onlineSavingsMsg'
Learn More

Toluene, OmniSolv, MilliporeSigma™

Supplier:  Technidata TX0737P1

 View more versions of this product

Catalog No. MTX0737P1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Toluene
108-88-3
0.87g/cm3 at 20°C
4 L
4.85°C (40.7°F), Closed cup
Aromatic
Liquid
C6H5CH3
29 hPa (20°C)
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
OmniSolv
-95.2°C
Colorless
110.4°C
1.2 to 8%(V)
535.85°C (996.5°F)
Poly-coated Glass Bottle
C7H8
MFCD00008512
UN 1294
Insoluble in water
CC1=CC=CC=C1
92.14
CHEBI:17578
≥99.9%

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 3, Packaging Group: II

Recommended Storage : Store at +15°C to +25°C

SDS
Documents

Documents

Promotions

Promotions