missing translation for 'onlineSavingsMsg'
Learn More

Toluene, CHROMASOLV™ Plus, for HPLC, Solstice

Catalog No. 6001707
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001707 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001707 Supplier Honeywell Inc Supplier No. 6505791L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Density 0.865g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Vapor Pressure 26 mmHg (25°C), 22 mmHg (20°C)
Linear Formula C6H5CH3
UN Number UN1294
Formula Weight 92.14g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Toluene
Show More Show Less
Hazard Statement P210-P261-P281-P301 + P310-P331
RTECSNumber XS5250000