missing translation for 'onlineSavingsMsg'
Learn More

Toluene, For pesticide residue analysis according to FDA, Honeywell Riedel-de Haën™

Catalog No. p-7072105
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001714 2.5 L
60046351 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001714 Supplier Honeywell Inc Supplier No. 344942.5L
Only null left

CHROMASOLV™, for pesticide residue analysis according to FDA

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Color Colorless
Density 0.865g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Linear Formula C6H5CH3
Vapor Pressure 22 mmHg (20°C), 26 mmHg (25°C)
UN Number UN1294
Beilstein 635760
Formula Weight 92.14g/mol
Percent Purity ≥99.7% (GC)
Grade CHROMASOLV
Chemical Name or Material Toluene
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
RTECSNumber XS5250000