missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.8%, Honeywell Burdick & Jackson
Specifications
Specifications
| Absorbance | 0.12 AU max. at 300 nm, 0.025 AU max. at 325 nm |
| CAS Max % | 100.0000% |
| Density | 0.867 g/cm3 |
| Boiling Point | 110.6°C |
| Quantity | 6 x 1 L |
| Flash Point | 4°C |
| Packaging | Glass Bottle |
| UN Number | UN1294 |
| Beilstein | 635760 |
| Formula Weight | 92.14 |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene |