missing translation for 'onlineSavingsMsg'
Learn More

Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA41841K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41841K2 1 L
AA41841AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41841K2 Supplier Thermo Scientific Chemicals Supplier No. 041841K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-88-3 | C7H8 | 92.14 g/mol

Toluene is widely used as an organic solvent in various industries like paints, pharmaceuticals, dyes, agrochemicals and polymers. It is used as an additive in gasoline and improves octane ratings, as a fullerene indicator, in paint thinners and silicone sealants, as a coolant in sodium cold traps used in nuclear reactor system loops and as a fuel for both two-stroke and four-stroke engines. It is also used as a raw material in the production of benzene, benzaldehyde, benzoic acid, toluene diisocyanate, benzyl-tert-butyl peroxide, xylene and many other chemical products. It plays an important role in biochemistry experiments to break open red blood cells in order to extract haemoglobin as well as used in the removal of cocaine from coca leaves to make coca-cola syrup. Further, it used as a standard material for chromatography and as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Density 0.867 g/mL
Quantity 1 L
Flash Point 4°C (40°F)
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range 99.8%
Beilstein 635760
Boiling Point 111°C
Solubility Information Miscible with ethanol,alcohol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Chemical Name or Material Toluene, anhydrous
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-625-9
RTECSNumber XS5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only